ContactUsLinkComponent
RABIF Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 143 amino acids (1-123 a.a.) and having a molecular mass of 16 kDa. The RABIF is fused to an 20 amino acid His Tag at N-Terminus and puri 1 * 10 µG
Catalog # BIRBORB81297-10
Supplier:  Biorbyt
RABIF Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 143 amino acids (1-123 a.a.) and having a molecular mass of 16 kDa. The RABIF is fused to an 20 amino acid His Tag at N-Terminus and puri 1 * 10 µG
Catalog # BIRBORB81297-10
Supplier:  Biorbyt