ContactUsLinkComponent
Contains 583 amino acid residues derived from the prototypical bovine serum albumin sequence. Suitable for use as biochemical, excip ient (an inert substance used as diluent or vehicle for protein), in culture media and chromatographic appli 1 * 10 g
Catalog # BIRBORB80972-10
Supplier:  Biorbyt
Contains 583 amino acid residues derived from the prototypical bovine serum albumin sequence. Suitable for use as biochemical, excip ient (an inert substance used as diluent or vehicle for protein), in culture media and chromatographic appli 1 * 10 g
Catalog # BIRBORB80972-10
Supplier:  Biorbyt