ContactUsLinkComponent
The active of Bivalirudin substance is synthetic 20 amino acid peptide. The amino acid sequence is Phe-Pro-Arg-Pro-Gly-Gly-Gly-Gly- Asn-Gly-Asp-Phe-Glu-Glu-Ile- Pro-Glu-Glu-Tyr-Leu. The Mw is 2180 dalton. 1 * 5 mg
Catalog # BIRBORB80969-5
Supplier:  Biorbyt
The active of Bivalirudin substance is synthetic 20 amino acid peptide. The amino acid sequence is Phe-Pro-Arg-Pro-Gly-Gly-Gly-Gly- Asn-Gly-Asp-Phe-Glu-Glu-Ile- Pro-Glu-Glu-Tyr-Leu. The Mw is 2180 dalton. 1 * 5 mg
Catalog # BIRBORB80969-5
Supplier:  Biorbyt