ContactUsLinkComponent
YWHAH Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 266 amino acids and having a molecular mass of 30.3 kDa. YWHAH is fused to N-Terminus His Tag and purified by proprietary chromatographic 1 * 25 µG
Catalog # BIRBORB80883-25
Supplier:  Biorbyt
YWHAH Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 266 amino acids and having a molecular mass of 30.3 kDa. YWHAH is fused to N-Terminus His Tag and purified by proprietary chromatographic 1 * 25 µG
Catalog # BIRBORB80883-25
Supplier:  Biorbyt