ContactUsLinkComponent
YWHAG Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 246 amino acids (2-247) and h aving a molecular mass of 36 kDa. YWHAG is fused to His Tag and purified by proprietary chromatographic tech 1 * 2 µG
  BIRBORB80882-2
 :  Biorbyt
YWHAG Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 246 amino acids (2-247) and h aving a molecular mass of 36 kDa. YWHAG is fused to His Tag and purified by proprietary chromatographic tech 1 * 2 µG
  BIRBORB80882-2
 :  Biorbyt
Frequently Bought Together