PRPS1 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 338 amino acids (1-318 a.a.) a nd having molecular wieght of 36.9kDa. The PRPS1 is is fused to 20 a.a. His-Tag at N-terminus and purified b 1 * 25 µG
Catalog # BIRBORB80865-25
Supplier: Biorbyt
Frequently Bought Together