ContactUsLinkComponent
MAP2K1 active Human Recombinant produced in Sf9 cells is glycosylated, polypeptide chain containing amino acids 2-393 having a molec ular mass of 47 kDa. MAP2K1 is fused to polyhistidine tag and is purified by proprietary chromatographic tec 1 * 5 µG
Catalog # BIRBORB80833-5
Supplier:  Biorbyt
MAP2K1 active Human Recombinant produced in Sf9 cells is glycosylated, polypeptide chain containing amino acids 2-393 having a molec ular mass of 47 kDa. MAP2K1 is fused to polyhistidine tag and is purified by proprietary chromatographic tec 1 * 5 µG
Catalog # BIRBORB80833-5
Supplier:  Biorbyt