ContactUsLinkComponent
Soluble LYVE1 Human Recombinant fused to GST-tag produced in E.coli is a single, non-glycosylated, polypeptide containing 243-323 am ino acids and having total molecular mass of 36 kDa. The LYVE-1 is purified by proprietary chromatographic t 1 * 2 µG
  BIRBORB80829-2
 :  Biorbyt
Soluble LYVE1 Human Recombinant fused to GST-tag produced in E.coli is a single, non-glycosylated, polypeptide containing 243-323 am ino acids and having total molecular mass of 36 kDa. The LYVE-1 is purified by proprietary chromatographic t 1 * 2 µG
  BIRBORB80829-2
 :  Biorbyt
Frequently Bought Together