ContactUsLinkComponent
ILK1 Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 452 amino acids fragment (1-45 2) having a molecular mass of 55.92kDa and fused with 4.5kDa amino-terminal hexahistidine tag. The ILK1 is p 1 * 10 µG
Catalog # BIRBORB80824-10
Supplier:  Biorbyt
ILK1 Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 452 amino acids fragment (1-45 2) having a molecular mass of 55.92kDa and fused with 4.5kDa amino-terminal hexahistidine tag. The ILK1 is p 1 * 10 µG
Catalog # BIRBORB80824-10
Supplier:  Biorbyt