ContactUsLinkComponent
HK2 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain fused to His tag at the N-terminal encodin g the sequence of 937 amino acids and having a molecular mass of 104.1 kDa.HXK2 is purified by proprietary c 1 * 10 µG
Catalog # BIRBORB80821-10
Supplier:  Biorbyt
HK2 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain fused to His tag at the N-terminal encodin g the sequence of 937 amino acids and having a molecular mass of 104.1 kDa.HXK2 is purified by proprietary c 1 * 10 µG
Catalog # BIRBORB80821-10
Supplier:  Biorbyt