ContactUsLinkComponent
Glucokinase Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain fused to His tag at the N-terminal encoding the sequence of 485 amino acids and having a molecular mass of 54.3 kDa.HK4 is purified by proprie 1 * 10 µG
Catalog # BIRBORB80818-10
Supplier:  Biorbyt
Glucokinase Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain fused to His tag at the N-terminal encoding the sequence of 485 amino acids and having a molecular mass of 54.3 kDa.HK4 is purified by proprie 1 * 10 µG
Catalog # BIRBORB80818-10
Supplier:  Biorbyt