GALK1 Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 412 amino acids (1-392 a.a.) a nd having a molecular mass of 44.4 kDa. The GALK1 is fused to 20 amino acid His-Tag at N-terminus and purifi 1 * 5 µG
Catalog # BIRBORB80817-5
Supplier: Biorbyt
Frequently Bought Together