ContactUsLinkComponent
IL19 Mouse Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 152 amino acids and having a mo lecular mass of 17722 Dalton. The IL-19 is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80468-2
Supplier:  Biorbyt
IL19 Mouse Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 152 amino acids and having a mo lecular mass of 17722 Dalton. The IL-19 is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80468-2
Supplier:  Biorbyt