ContactUsLinkComponent
NGF-beta Human Recombinant produced in CHO is homodimer, glycosylated, polypeptide chain containing identical 120 amino acids and ha ving a molecular mass of 16,950 Dalton.The NGF-b is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB80280-20
Supplier:  Biorbyt
NGF-beta Human Recombinant produced in CHO is homodimer, glycosylated, polypeptide chain containing identical 120 amino acids and ha ving a molecular mass of 16,950 Dalton.The NGF-b is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB80280-20
Supplier:  Biorbyt