ContactUsLinkComponent
NGF beta Mouse produced in Submaxillary Gland of Grown Mouse is homodimer, non-glycosylated, polypeptide chain containing identical 120 amino acids and having a molecular mass of 13,471 Dalton each.The NGF beta Mouse is purified by advanced 1 * 5 µG
Catalog # BIRBORB80282-5
Supplier:  Biorbyt
NGF beta Mouse produced in Submaxillary Gland of Grown Mouse is homodimer, non-glycosylated, polypeptide chain containing identical 120 amino acids and having a molecular mass of 13,471 Dalton each.The NGF beta Mouse is purified by advanced 1 * 5 µG
Catalog # BIRBORB80282-5
Supplier:  Biorbyt