ContactUsLinkComponent
Hexarelin stimulates GH secretion. Hexarelin is more resistant to proteolytic degradation than GHRP-6. Hexarelin has amino acids H-H is-D-2-Methyl-Trp-Ala-Trp-D-Phe-Lys-NH2 and having molecular weight of 887 Dalton. The Molecular Formula is 1 * 20 mg
Catalog # BIRBORB80732-20
Supplier:  Biorbyt
Hexarelin stimulates GH secretion. Hexarelin is more resistant to proteolytic degradation than GHRP-6. Hexarelin has amino acids H-H is-D-2-Methyl-Trp-Ala-Trp-D-Phe-Lys-NH2 and having molecular weight of 887 Dalton. The Molecular Formula is 1 * 20 mg
Catalog # BIRBORB80732-20
Supplier:  Biorbyt