ContactUsLinkComponent
IP-10 His Tag Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 77 amino acids fragme nt (22-98) and having total molecular mass of 8.5 kDa with an amino-terminal hexahistidine tag. The IP-10 Hi 1 * 5 µG
Catalog # BIRBORB80668-5
Supplier:  Biorbyt
IP-10 His Tag Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 77 amino acids fragme nt (22-98) and having total molecular mass of 8.5 kDa with an amino-terminal hexahistidine tag. The IP-10 Hi 1 * 5 µG
Catalog # BIRBORB80668-5
Supplier:  Biorbyt