ContactUsLinkComponent
IL-17A/F Rat Recombinant produced in E.coli is heterodimeric, non-glycosylated polypeptide chain containing monomeric subunit of eac h IL-17A IL-17F. The dimer contains 271 amino acids and having total molecular mass of 30.7 kDa. The IL-17A/ 1 * 100 µG
Catalog # BIRBORB80459-100
Supplier:  Biorbyt
IL-17A/F Rat Recombinant produced in E.coli is heterodimeric, non-glycosylated polypeptide chain containing monomeric subunit of eac h IL-17A IL-17F. The dimer contains 271 amino acids and having total molecular mass of 30.7 kDa. The IL-17A/ 1 * 100 µG
Catalog # BIRBORB80459-100
Supplier:  Biorbyt