ContactUsLinkComponent
IL17 Murine Recombinant produced in E.coli is homodimeric, non-glycosylated polypeptide chain containing total of 266 amino acids an d having a molecular mass of 29,956 Dalton. The IL-17 is purified by proprietary chromatographic techniques. 1 * 25 µG
  BIRBORB80455-25
 :  Biorbyt
IL17 Murine Recombinant produced in E.coli is homodimeric, non-glycosylated polypeptide chain containing total of 266 amino acids an d having a molecular mass of 29,956 Dalton. The IL-17 is purified by proprietary chromatographic techniques. 1 * 25 µG
  BIRBORB80455-25
 :  Biorbyt
Frequently Bought Together