ContactUsLinkComponent
IL-17A/F Human Recombinant produced in E.coli is heterodimeric, non-glycosylated polypeptide chain containing monomeric subunit of e ach IL-17A IL-17F. The active dimer contains 271 amino acids and having total molecular mass of 30.7 kDa. Th 1 * 10 µG
  BIRBORB80454-10
 :  Biorbyt
IL-17A/F Human Recombinant produced in E.coli is heterodimeric, non-glycosylated polypeptide chain containing monomeric subunit of e ach IL-17A IL-17F. The active dimer contains 271 amino acids and having total molecular mass of 30.7 kDa. Th 1 * 10 µG
  BIRBORB80454-10
 :  Biorbyt
Frequently Bought Together