ContactUsLinkComponent
IL17E Human Recombinant produced in E.coli is homodimeric, non-glycosylated polypeptide chain containing 146 amino acids and having a molecular mass of 33.7 kDa. The IL-25 Human is purified by proprietary chromatographic techniques. 1 * 25 µG
Catalog # BIRBORB80453-25
Supplier:  Biorbyt
IL17E Human Recombinant produced in E.coli is homodimeric, non-glycosylated polypeptide chain containing 146 amino acids and having a molecular mass of 33.7 kDa. The IL-25 Human is purified by proprietary chromatographic techniques. 1 * 25 µG
Catalog # BIRBORB80453-25
Supplier:  Biorbyt