ContactUsLinkComponent
IL17A Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 132 amino acids fragment (24- 155) having molecular weight of 21.5kDa and fused with 4.5kDa amino-terminal hexahistidine tag. The IL-17A H 1 * 10 µG
Catalog # BIRBORB80452-10
Supplier:  Biorbyt
IL17A Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 132 amino acids fragment (24- 155) having molecular weight of 21.5kDa and fused with 4.5kDa amino-terminal hexahistidine tag. The IL-17A H 1 * 10 µG
Catalog # BIRBORB80452-10
Supplier:  Biorbyt