ContactUsLinkComponent
IL16 Mouse Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 127 amino acids and having a mo lecular mass of 13.2 kDa. The Mouse IL-16 is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80451-10
Supplier:  Biorbyt
IL16 Mouse Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 127 amino acids and having a mo lecular mass of 13.2 kDa. The Mouse IL-16 is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80451-10
Supplier:  Biorbyt