ContactUsLinkComponent
IL16 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 130 amino acids and having a mo lecular mass of 13.5 kDa. The IL-16 is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80449-2
Supplier:  Biorbyt
IL16 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 130 amino acids and having a mo lecular mass of 13.5 kDa. The IL-16 is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80449-2
Supplier:  Biorbyt