ContactUsLinkComponent
IL15 Porcine Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 114 amino acids and having a molecular mass of 13,118 Dalton. The IL-15 is purified by proprietary chromatographic techniques. 1 * 2 µG
  BIRBORB80448-2
 :  Biorbyt
IL15 Porcine Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 114 amino acids and having a molecular mass of 13,118 Dalton. The IL-15 is purified by proprietary chromatographic techniques. 1 * 2 µG
  BIRBORB80448-2
 :  Biorbyt
Frequently Bought Together