ContactUsLinkComponent
IL15 Mouse Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 152 amino acids (49-162 a.a.) a nd having a molecular mass of 17.6 kDa. The IL-15 is fused to 37 amino acid His Tag at N-terminus and purifi 1 * 10 µG
Catalog # BIRBORB80446-10
Supplier:  Biorbyt
IL15 Mouse Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 152 amino acids (49-162 a.a.) a nd having a molecular mass of 17.6 kDa. The IL-15 is fused to 37 amino acid His Tag at N-terminus and purifi 1 * 10 µG
Catalog # BIRBORB80446-10
Supplier:  Biorbyt