ContactUsLinkComponent
IL12 p40 His Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 306 amino acids fragme nt (23-328) with an amino-terminal hexahistidine tag. The IL-12 p40 His is purified by proprietary chromatog 1 * 2 µG
Catalog # BIRBORB80441-2
Supplier:  Biorbyt
IL12 p40 His Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 306 amino acids fragme nt (23-328) with an amino-terminal hexahistidine tag. The IL-12 p40 His is purified by proprietary chromatog 1 * 2 µG
Catalog # BIRBORB80441-2
Supplier:  Biorbyt