ContactUsLinkComponent
IL12 p35 His Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 197 amino acids fragme nt (57-253) with an amino-terminal hexahistidine tag. The IL-12 p35 His is purified by proprietary chromatog 1 * 10 µG
Catalog # BIRBORB80440-10
Supplier:  Biorbyt
IL12 p35 His Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 197 amino acids fragme nt (57-253) with an amino-terminal hexahistidine tag. The IL-12 p35 His is purified by proprietary chromatog 1 * 10 µG
Catalog # BIRBORB80440-10
Supplier:  Biorbyt