ContactUsLinkComponent
IL12 Rat Recombinant produced in Sf9 insect cells is glycosylated disulfide linked heterodimeric polypeptide containing 503 amino ac ids and having molecular weight of 70 kDa comprised of disulfide-bonded 35 kDa (p35) and 40 kDa (p40) subuni 1 * 10 µG
Catalog # BIRBORB80439-10
Supplier:  Biorbyt
IL12 Rat Recombinant produced in Sf9 insect cells is glycosylated disulfide linked heterodimeric polypeptide containing 503 amino ac ids and having molecular weight of 70 kDa comprised of disulfide-bonded 35 kDa (p35) and 40 kDa (p40) subuni 1 * 10 µG
Catalog # BIRBORB80439-10
Supplier:  Biorbyt