ContactUsLinkComponent
Human IL9 Recombinant produced in E.coli is a single, non-glycosylated single polypeptide chain containing 127amino acids and having a molecular mass of 14,004 Dalton. The IL-9 is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80435-10
Supplier:  Biorbyt
Human IL9 Recombinant produced in E.coli is a single, non-glycosylated single polypeptide chain containing 127amino acids and having a molecular mass of 14,004 Dalton. The IL-9 is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80435-10
Supplier:  Biorbyt