ContactUsLinkComponent
MICB Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide containing 326 amino acids and having a molecul ar mass of 37kDa. The sequence contains the extracellular domain of the mature human MICB (amino acid residu 1 * 10 µG
Catalog # BIRBORB80177-10
Supplier:  Biorbyt
MICB Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide containing 326 amino acids and having a molecul ar mass of 37kDa. The sequence contains the extracellular domain of the mature human MICB (amino acid residu 1 * 10 µG
Catalog # BIRBORB80177-10
Supplier:  Biorbyt