Exenatide is a single, non-glycosylated, peptide containing 39 amino acids and having a molecular mass of 4186.6 Dalton. Exenatide h as the empirical formula C184H282N50O60S. 1 * 10 mg
Catalog # BIRBORB80712-10
Supplier: Biorbyt
Frequently Bought Together