ContactUsLinkComponent
TNFR1 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 162 amino acids and having to tal molecular mass of 18.2 kDa. TNFR1 Human Recombinant is purified by proprietary chromatographic technique 1 * 20 µG
Katalogové číslo BIRBORB80623-20
Dodavatel:  Biorbyt
TNFR1 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 162 amino acids and having to tal molecular mass of 18.2 kDa. TNFR1 Human Recombinant is purified by proprietary chromatographic technique 1 * 20 µG
Katalogové číslo BIRBORB80623-20
Dodavatel:  Biorbyt
Související informace
Frequently Bought Together