ContactUsLinkComponent
4-1BB Soluble Receptor Recombinant Human also called TNF receptor superfamily member produced in E.coli is a single, non-glycosylate d polypeptide chain containing 167 amino acids, having a molecular mass of 17718 Dalton and containing the c 1 * 20 µG
  BIRBORB80617-20
 :  Biorbyt
4-1BB Soluble Receptor Recombinant Human also called TNF receptor superfamily member produced in E.coli is a single, non-glycosylate d polypeptide chain containing 167 amino acids, having a molecular mass of 17718 Dalton and containing the c 1 * 20 µG
  BIRBORB80617-20
 :  Biorbyt
Frequently Bought Together