ContactUsLinkComponent
Human IL3 Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 133 amino acids fragment (20-15 2) and having total molecular mass of 19.59kDa with an amino-terminal hexahistidine tag. The IL3 His is puri 1 * 10 µG
Catalog # BIRBORB80419-10
Supplier:  Biorbyt
Human IL3 Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 133 amino acids fragment (20-15 2) and having total molecular mass of 19.59kDa with an amino-terminal hexahistidine tag. The IL3 His is puri 1 * 10 µG
Catalog # BIRBORB80419-10
Supplier:  Biorbyt