ContactUsLinkComponent
Soluble FGFR-4a (IIIc) Fc Chimera Human Recombinant fused with Xa cleavage site with the Fc part of human IgG1 produced in baculovir us is heterodimeric, glycosylated, Polypeptide chain and having a molecular mass of 170 kDa. The FGFR4 is pu 1 * 10 µG
Catalog # BIRBORB80806-10
Supplier:  Biorbyt
Soluble FGFR-4a (IIIc) Fc Chimera Human Recombinant fused with Xa cleavage site with the Fc part of human IgG1 produced in baculovir us is heterodimeric, glycosylated, Polypeptide chain and having a molecular mass of 170 kDa. The FGFR4 is pu 1 * 10 µG
Catalog # BIRBORB80806-10
Supplier:  Biorbyt
Frequently Bought Together