ContactUsLinkComponent
Desmopressin also called ADH (Anti-Diuretic Hormone) has molecular formula of C46H64N14O12S2, Mpr-Tyr-Phe-Gln-Asn-Cys-Pro-D-Arg-Gly- NH2 having Mw of 1069.23 Dalton, is the first vasopressin analog with very high and very specific antidiuret 1 * 25 mg
Catalog # BIRBORB80706-25
Supplier:  Biorbyt
Desmopressin also called ADH (Anti-Diuretic Hormone) has molecular formula of C46H64N14O12S2, Mpr-Tyr-Phe-Gln-Asn-Cys-Pro-D-Arg-Gly- NH2 having Mw of 1069.23 Dalton, is the first vasopressin analog with very high and very specific antidiuret 1 * 25 mg
Catalog # BIRBORB80706-25
Supplier:  Biorbyt