ContactUsLinkComponent
TGF-b Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 113 amino acids (279-390 a.a. ) and having total molecular mass of 12.9 kDa. TGF-b (113 a.a.) is purified by proprietary chromatographic t 1 * 10 µG
Catalog # BIRBORB80611-10
Supplier:  Biorbyt
TGF-b Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 113 amino acids (279-390 a.a. ) and having total molecular mass of 12.9 kDa. TGF-b (113 a.a.) is purified by proprietary chromatographic t 1 * 10 µG
Catalog # BIRBORB80611-10
Supplier:  Biorbyt