ContactUsLinkComponent
TFF-2 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 116 amino acids (24-129) whic h includes 10 amino acid His Tag fused at N-terminus and having total molecular mass of 13.2 kDa. TFF2 Human 1 * 20 µG
Catalog # BIRBORB80606-20
Supplier:  Biorbyt
TFF-2 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 116 amino acids (24-129) whic h includes 10 amino acid His Tag fused at N-terminus and having total molecular mass of 13.2 kDa. TFF2 Human 1 * 20 µG
Catalog # BIRBORB80606-20
Supplier:  Biorbyt