ContactUsLinkComponent
TFF-1 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 70 amino acids (25-84) which includes 10 amino acid His Tag and having total molecular mass of 7.9 kDa. TFF-1 Human Recombinant is purifi 1 * 20 µG
Catalog # BIRBORB80605-20
Supplier:  Biorbyt
TFF-1 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 70 amino acids (25-84) which includes 10 amino acid His Tag and having total molecular mass of 7.9 kDa. TFF-1 Human Recombinant is purifi 1 * 20 µG
Catalog # BIRBORB80605-20
Supplier:  Biorbyt