ContactUsLinkComponent
TDGF1 Human Recombinant produced in HEK-293 cells is a single, glycosylated, polypeptide chain containing 139 amino acids with molec ular weight of 17.8 kDa though on SDS-PAGE migrates at about 25kDa due to the glycosilation. The TDGF1 is pu 1 * 10 µG
Catalog # BIRBORB80604-10
Supplier:  Biorbyt
TDGF1 Human Recombinant produced in HEK-293 cells is a single, glycosylated, polypeptide chain containing 139 amino acids with molec ular weight of 17.8 kDa though on SDS-PAGE migrates at about 25kDa due to the glycosilation. The TDGF1 is pu 1 * 10 µG
Catalog # BIRBORB80604-10
Supplier:  Biorbyt