ContactUsLinkComponent
Interleukin2 Porcine Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 134 amino acids and h aving a molecular mass of 15217 Dalton. The IL-2 is purified by proprietary chromatographic techniques. 1 * 10 µG
  BIRBORB80416-10
 :  Biorbyt
Interleukin2 Porcine Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 134 amino acids and h aving a molecular mass of 15217 Dalton. The IL-2 is purified by proprietary chromatographic techniques. 1 * 10 µG
  BIRBORB80416-10
 :  Biorbyt
Frequently Bought Together