ContactUsLinkComponent
Human IL2 Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 133 amino acids fragment (21-15 3) having molecular weight of 20kDa and fused with 4.5kDa amino-terminal hexahistidine tag. The IL-2 His is 1 * 10 µG
Catalog # BIRBORB80413-10
Supplier:  Biorbyt
Human IL2 Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 133 amino acids fragment (21-15 3) having molecular weight of 20kDa and fused with 4.5kDa amino-terminal hexahistidine tag. The IL-2 His is 1 * 10 µG
Catalog # BIRBORB80413-10
Supplier:  Biorbyt