ContactUsLinkComponent
ST2 Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 310 amino acids fragment (19-32 8) corresponding to the soluble ST2 coding sequence, having molecular weight of 36kDa and fused with 4.5kDa 1 * 10 µG
Catalog # BIRBORB80411-10
Supplier:  Biorbyt
ST2 Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 310 amino acids fragment (19-32 8) corresponding to the soluble ST2 coding sequence, having molecular weight of 36kDa and fused with 4.5kDa 1 * 10 µG
Catalog # BIRBORB80411-10
Supplier:  Biorbyt