ContactUsLinkComponent
Human IL1 beta His-tag Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 153 amino acids fr agment (117-269) and having total molecular mass of 21.88 kDa with an amino-terminal hexahistidine tag. The 1 * 5 µG
Catalog # BIRBORB80407-5
Supplier:  Biorbyt
Human IL1 beta His-tag Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 153 amino acids fr agment (117-269) and having total molecular mass of 21.88 kDa with an amino-terminal hexahistidine tag. The 1 * 5 µG
Catalog # BIRBORB80407-5
Supplier:  Biorbyt