ContactUsLinkComponent
SAA1 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 125 amino acids (19-122 a.a.) and having total molecular mass of 13.9 kDa. SAA1 is fused to 20 amino acid His Tag at N-terminus and purifi 1 * 10 µG
Catalog # BIRBORB80598-10
Supplier:  Biorbyt
SAA1 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 125 amino acids (19-122 a.a.) and having total molecular mass of 13.9 kDa. SAA1 is fused to 20 amino acid His Tag at N-terminus and purifi 1 * 10 µG
Catalog # BIRBORB80598-10
Supplier:  Biorbyt