ContactUsLinkComponent
Insulin-Like Growth Factor-II Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 67 am ino acids and having a molecular mass of 7505 Dalton. IGF-II is purified by proprietary chromatographic tech 1 * 50 µG
  BIRBORB80399-50
 :  Biorbyt
Insulin-Like Growth Factor-II Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 67 am ino acids and having a molecular mass of 7505 Dalton. IGF-II is purified by proprietary chromatographic tech 1 * 50 µG
  BIRBORB80399-50
 :  Biorbyt
Frequently Bought Together