ContactUsLinkComponent
Insulin-Like Growth Factor-II Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 67 am ino acids and having a molecular mass of 7505 Dalton. IGF-II is purified by proprietary chromatographic tech 1 * 10 µG
Catalog # BIRBORB80399-10
Supplier:  Biorbyt
Insulin-Like Growth Factor-II Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 67 am ino acids and having a molecular mass of 7505 Dalton. IGF-II is purified by proprietary chromatographic tech 1 * 10 µG
Catalog # BIRBORB80399-10
Supplier:  Biorbyt