ContactUsLinkComponent
IRF-3 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide containing 111 amino acids (1-112) and having a molecular mass of 13 kDa.The Interferon Regulatory Factor-3 is purified by proprietary chromatographic te 1 * 20 µG
Catalog # BIRBORB80395-20
Supplier:  Biorbyt
IRF-3 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide containing 111 amino acids (1-112) and having a molecular mass of 13 kDa.The Interferon Regulatory Factor-3 is purified by proprietary chromatographic te 1 * 20 µG
Catalog # BIRBORB80395-20
Supplier:  Biorbyt