ContactUsLinkComponent
Interferon-g Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 143 amino acids fragme nt (24-166) and having a molecular mass of 21.28 kDa with an amino-terminal hexahistidine tag. The IFN-g-His 1 * 10 µG
Catalog # BIRBORB80391-10
Supplier:  Biorbyt
Interferon-g Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 143 amino acids fragme nt (24-166) and having a molecular mass of 21.28 kDa with an amino-terminal hexahistidine tag. The IFN-g-His 1 * 10 µG
Catalog # BIRBORB80391-10
Supplier:  Biorbyt